ADVH7F37F679 AChemBlock

1-(azetidin-3-yl)hydrazine dihydrochloride

  • MDL number MFCD22380550
  • Cas Number : 1305282-79-4
  • Molecular Weight : 160.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-azetidin-3-yl-hydrazine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29510
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart