ADVH7F37F67D AChemBlock

3-(2-aminopyridin-4-yl)propanoic acid

  • MDL number MFCD22412893
  • Cas Number : 751456-76-5
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-2-aminopyridin-4-yl-propanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29533
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart