ADVH7F37F67E AChemBlock

3-(6-fluoropyridin-3-yl)propanoic acid

  • MDL number MFCD18803196
  • Cas Number : 944998-15-6
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-6-fluoropyridin-3-yl-propanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29534
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart