ADVH7F37F682 AChemBlock

N-(3,4-dimethylbenzyl)cyclopropanamine

  • MDL number MFCD11204719
  • Cas Number : 936023-14-2
  • Molecular Weight : 175.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-3-4-dimethylbenzyl-cyclopropanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29553
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart