ADVH7F37F68A AChemBlock

2-chloro-1-(4-chloropyridin-2-yl)ethanone

  • MDL number MFCD22380407
  • Cas Number : 1357946-32-7
  • Molecular Weight : 190.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-1-4-chloropyridin-2-yl-ethanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29576
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart