ADVH7F37F68D AChemBlock

5-tert-butyl-2`,3`,5`,6`-tetramethylbiphenyl-2-ol

  • MDL number MFCD22689773
  • Cas Number : 1416713-98-8
  • Molecular Weight : 282.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butyl-2-3-5-6-tetramethylbiphenyl-2-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29580
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart