ADVH7F37F6AC AChemBlock

2-methyl-5-(pyridin-4-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD24501065
  • Cas Number : 1248408-55-0
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-5-pyridin-4-yl-1h-pyrrole-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29659
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart