ADVH7F37F6B1 AChemBlock

tert-butyl 4-(5-aminopyridin-2-yl)piperidine-1-carboxylate

  • MDL number MFCD08437656
  • Cas Number : 885693-48-1
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-5-aminopyridin-2-yl-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29667
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart