ADVH7F37F6BA AChemBlock

2-methylbenzo[d]thiazol-4-ol

  • MDL number MFCD11656604
  • Cas Number : 94977-59-0
  • Molecular Weight : 165.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methylbenzo-d-thiazol-4-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29691
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart