ADVH7F37F6C0 AChemBlock

2-amino-1-(3,4-dimethoxyphenyl)propan-1-one hydrochloride

  • MDL number MFCD04973548
  • Cas Number : 90253-98-8
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-amino-1-3-4-dimethoxyphenyl-propan-1-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29703
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart