ADVH7F37F6C2 AChemBlock

methyl thieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD18833774
  • Cas Number : 28783-21-3
  • Molecular Weight : 193.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-thieno-2-3-c-pyridine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29705
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart