ADVH7F37F6C6 AChemBlock

5-chloroH-imidazo[1,2-a]pyridin-8-amine

  • MDL number MFCD22013194
  • Cas Number : 1357947-17-1
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-chloroh-imidazo-1-2-a-pyridin-8-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29718
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart