ADVH7F37F6C8 AChemBlock

1-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl-3-boronic acid

  • MDL number MFCD10696651
  • Cas Number : 521985-24-0
  • Molecular Weight : 175.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-pyrrolo-2-3-b-pyridin-3-yl-3-boronic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29729
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart