ADVH7F37F6E6 AChemBlock

piperidine-3-sulfonamide hydrochloride

  • MDL number MFCD11099460
  • Cas Number : 1170558-67-4
  • Molecular Weight : 200.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/piperidine-3-sulfonamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29812
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart