ADVH7F37F6EA AChemBlock

tert-butyl 6-chloro-4-formylpyridin-3-ylcarbamate

  • MDL number MFCD26403907
  • Cas Number : 1238324-67-8
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-chloro-4-formylpyridin-3-ylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29820
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart