ADVH7F37F6FB AChemBlock

4-chloro-1H-indazole-3-carboxamide

  • MDL number MFCD24447530
  • Cas Number : 1432754-19-2
  • Molecular Weight : 195.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-1h-indazole-3-carboxamide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29865
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart