ADVH7F37F703 AChemBlock

6-(naphthalen-2-yloxy)pyridin-3-amine

  • MDL number MFCD09734043
  • Cas Number : 83414-50-0
  • Molecular Weight : 236.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-naphthalen-2-yloxy-pyridin-3-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29889
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart