ADVH7F37F705 AChemBlock

7-bromo-1,2,3,4-tetrahydro-3-methylisoquinoline

  • MDL number MFCD11559077
  • Cas Number : 848185-12-6
  • Molecular Weight : 226.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1-2-3-4-tetrahydro-3-methylisoquinoline.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29894
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart