ADVH7F37F711 AChemBlock

6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

  • MDL number MFCD23701113
  • Cas Number : 425702-95-0
  • Molecular Weight : 203.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethyl-1-2-4-triazolo-4-3-a-pyridin-3-2h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O29935
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart