ADVH7F37F737 AChemBlock

2-methyl-2-phenylcyclohexanone

  • MDL number MFCD00230846
  • Cas Number : 17206-54-1
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-2-phenylcyclohexanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30049
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart