ADVH7F37F741 AChemBlock

2,4,6-trichlorobenzenethiol

  • MDL number MFCD00052937
  • Cas Number : 24207-66-7
  • Molecular Weight : 213.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-6-trichlorobenzenethiol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30066
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart