ADVH7F37F753 AChemBlock

(2-chloro-5-(trifluoromethyl)pyridin-3-yl)methanol

  • MDL number MFCD11848356
  • Cas Number : 943551-28-8
  • Molecular Weight : 211.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-trifluoromethyl-pyridin-3-yl-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30112
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart