ADVH7F37F774 AChemBlock

6-bromobenzo[d][1,3]dioxole-4-carbaldehyde

  • MDL number MFCD04037050
  • Cas Number : 72744-55-9
  • Molecular Weight : 229.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-bromobenzo-d-1-3-dioxole-4-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30185
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart