ADVH7F37F788 AChemBlock

4-isothiocyanato-1-methyl-1H-pyrazole

  • MDL number MFCD04969718
  • Cas Number : 1001500-53-3
  • Molecular Weight : 139.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-isothiocyanato-1-methyl-1h-pyrazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30226
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart