ADVH7F37F795 AChemBlock

(E)-ethyl 3-(2-amino-5-bromophenyl)acrylate

  • MDL number MFCD14155808
  • Cas Number : 950759-54-3
  • Molecular Weight : 270.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/e-ethyl-3-2-amino-5-bromophenyl-acrylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30269
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart