ADVH7F37F7CF AChemBlock

(benzo[b]thiophen-7-yl)methanamine hydrochloride

  • MDL number MFCD27924152
  • Cas Number : 870562-96-2
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/benzo-b-thiophen-7-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30410
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart