ADVH7F37F8E0 AChemBlock

6-chloro-2-methyl-3H-imidazo[4,5-c]pyridine

  • MDL number MFCD18256899
  • Cas Number : 7205-42-7
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-2-methyl-3h-imidazo-4-5-c-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30942
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart