ADVH7F37F8F5 AChemBlock

7-Bromoimidazo[1,2-a]pyridin-8-amine

  • MDL number MFCD22199338
  • Cas Number : 1357945-44-8
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-bromoimidazo-1-2-a-pyridin-8-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O30969
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart