ADVH7F37F910 AChemBlock

3-chloro-4-ethynylpyridine

  • MDL number MFCD13189773
  • Cas Number : 1196156-71-4
  • Molecular Weight : 137.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/3-chloro-4-ethynylpyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=O31010
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart