ADVH7F37F924 AChemBlock

1H-pyrrolo[2,3-c]pyridin-3-amine dihydrochloride

  • MDL number MFCD16628054
  • Cas Number : 1257535-55-9
  • Molecular Weight : 206.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrrolo-2-3-c-pyridin-3-amine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31035
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart