ADVH7F37F932 AChemBlock

ethyl 4-(benzyloxy)-1H-indole-2-carboxylate

  • MDL number MFCD00016687
  • Cas Number : 27737-55-9
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-benzyloxy-1h-indole-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31054
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart