ADVH7F37F945 AChemBlock

N-((2-chlorothiazol-5-yl)methyl)ethanamine

  • MDL number MFCD18837199
  • Cas Number : 120740-07-0
  • Molecular Weight : 176.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-2-chlorothiazol-5-yl-methyl-ethanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31076
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart