ADVH7F37F94B AChemBlock

3-(2-(piperidin-1-yl)ethoxy)benzenamine

  • MDL number MFCD09035171
  • Cas Number : 373824-23-8
  • Molecular Weight : 220.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-2-piperidin-1-yl-ethoxy-benzenamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31086
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart