ADVH7F37F9A6 AChemBlock

4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(methylsulfonyl)pyridine

  • MDL number MFCD18727629
  • Cas Number : 1353745-98-8
  • Molecular Weight : 297.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-methyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-2-methylsulfonyl-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31211
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart