ADVH7F37F9B1 AChemBlock

1,4-dibromobut-2-yne

  • MDL number MFCD00039197
  • Cas Number : 2219-66-1
  • Molecular Weight : 211.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-4-dibromobut-2-yne.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31222
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart