ADVH7F37F9B6 AChemBlock

3-(chloromethyl)-2-isopropoxypyridine

  • MDL number MFCD14607210
  • Cas Number : 1248614-20-1
  • Molecular Weight : 185.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-chloromethyl-2-isopropoxypyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31228
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart