ADVH7F37F9BA AChemBlock

3-(4-bromophenoxy)propane-1,2-diol

  • MDL number MFCD01698162
  • Cas Number : 63834-59-3
  • Molecular Weight : 247.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-4-bromophenoxy-propane-1-2-diol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31232
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart