ADVH7F37F9BC AChemBlock

2-(2-ethoxyethoxy)-5-bromopyridine

  • MDL number MFCD14652079
  • Cas Number : 1249442-19-0
  • Molecular Weight : 246.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-ethoxyethoxy-5-bromopyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31234
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart