ADVH7F37F9BE AChemBlock

methyl 4-chloro-5-cyclopropyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD16103386
  • Cas Number : 1291177-21-3
  • Molecular Weight : 200.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-chloro-5-cyclopropyl-1h-pyrazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31236
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart