ADVH7F37F9C3 AChemBlock

5-(ethoxycarbonyl)furan-2-carboxylic acid

  • MDL number MFCD11045350
  • Cas Number : 32933-01-0
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-ethoxycarbonyl-furan-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31242
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart