ADVH7F37F9CD AChemBlock

tert-butyl 4-((1,3-dimethyl-1H-pyrazol-4-yl)methyl)piperazine-1-carboxylate

  • MDL number MFCD28656710
  • Cas Number : 1951444-93-1
  • Molecular Weight : 294.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/tert-butyl-4-1-3-dimethyl-1h-pyrazol-4-yl-methyl-piperazine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/ac_sds?Catalog=O31253
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart