ADVH7F37F9D2 AChemBlock

7-bromothiochroman-4-one

  • MDL number MFCD22426388
  • Cas Number : 13735-16-5
  • Molecular Weight : 243.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-bromothiochroman-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31259
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart