ADVH7F37F9D3 AChemBlock

2-(4-(trifluoromethyl)phenyl)morpholine

  • MDL number MFCD11646253
  • Cas Number : 62243-72-5
  • Molecular Weight : 231.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-trifluoromethyl-phenyl-morpholine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31260
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart