ADVH7F37F9E2 AChemBlock

(2-chloro-5-methylpyridin-3-yl)methanamine hydrochloride

  • MDL number MFCD24447566
  • Cas Number : 1432754-65-8
  • Molecular Weight : 193.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-methylpyridin-3-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31277
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart