ADVH7F37F9E4 AChemBlock

tert-butyl 2-chloro-5-methylpyridin-4-ylcarbamate

  • MDL number MFCD26936365
  • Cas Number : 1820717-60-9
  • Molecular Weight : 242.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-chloro-5-methylpyridin-4-ylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31279
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart