ADVH7F37F9E8 AChemBlock

8-(prop-2-ynyl)-8-azaspiro[4.5]decane-7,9-dione

  • MDL number MFCD14631782
  • Cas Number : 25032-23-9
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-prop-2-ynyl-8-azaspiro-4-5-decane-7-9-dione.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31283
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart