ADVH7F37F9F8 AChemBlock

(3,3,5,5-tetramethylcyclohexyl)methanamine hydrochloride

  • MDL number MFCD28656565
  • Cas Number : 219835-66-2
  • Molecular Weight : 205.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3-5-5-tetramethylcyclohexyl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31300
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart