ADVH7F37F9FD AChemBlock

5-(4-methoxybenzyloxy)-2-ethynyl-1,3-difluorobenzene

  • MDL number MFCD24447564
  • Cas Number : 1432754-28-3
  • Molecular Weight : 274.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-4-methoxybenzyloxy-2-ethynyl-1-3-difluorobenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31305
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart