ADVH7F37FA00 AChemBlock

2-(trifluoromethyl)-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD18726737
  • Cas Number : 1612287-99-6
  • Molecular Weight : 287.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-4-methyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31308
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart