ADVH7F37FA0F AChemBlock

6-(trifluoromethyl)-4-hydroxypyridine-3-carboxylic acid

  • MDL number MFCD13188821
  • Cas Number : 1060810-79-8
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethyl-4-hydroxypyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=O31325
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart